missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1H Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526102
Code : LD
Additional Details : Weight : 0.01000kg
Description
PPM1H Polyclonal antibody specifically detects PPM1H in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PPM1H | |
Polyclonal | |
Unconjugated | |
PPM1H | |
A430075L18Rik; ARHCL1; C030002B11Rik; KIAA1157; NERPP-2C; neurite extension-related protein phosphatase related to PP2C; Ppm1h; Protein phosphatase 1H; protein phosphatase 1H (PP2C domain containing); protein phosphatase, Mg2+/Mn2+ dependent 1H; protein phosphatase, Mg2+/Mn2+ dependent, 1H; ras homolog gene family, member C like 1; URCC2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
319468, 57460 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Q3UYC0, Q9ULR3 | |
PPM1H | |
KLH conjugated synthetic peptide between 235-263 amino acids from the central region of human PPM1H | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |