Learn More
Cyclophilin E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541808
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KKFSGKTLEE NKEEEGSEPP KAETQEGEPI AKKARSNPQV YMDIKIGNKP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Sheep: 100%; Yeast: 100%
Cyclophilin E Polyclonal antibody specifically detects Cyclophilin E in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Cyclophilin E | |
Polyclonal | |
Unconjugated | |
PPIE | |
2010010D16Rik; Ab1-210; Cyclophilin E; cyclophilin-33; CYP33; CYP-33; Peptidyl-prolyl cis-trans isomerase E; peptidylprolyl isomerase E; peptidylprolyl isomerase E (cyclophilin E); PPIase E; PPIE; Rotamase E | |
Rabbit | |
Protein A | |
RUO | |
10450 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UNP9 | |
PPIE | |
Synthetic peptide directed towards the middle region of human PPIE | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |