missing translation for 'onlineSavingsMsg'
Learn More

PP2A Catalytic Subunit Rat anti-Human, Clone: 234532, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB16531

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448805

  • £277.83 / 100µg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PP2A Catalytic Subunit Monoclonal antibody MAB16531 Specifically detects PP2A Catalytic Subunit in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

PP2A alpha
234532
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PPP2CA
E. coli-derived recombinant human PP2A-alpha Catalytic Subunit Trp209-Leu309 Accession # P67775
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5515
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
P67775
EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, PPP2CA, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human PP2A Catalytic Subunit in direct ELISAs. Because PP2A-alpha and PP2A-beta Catalytic Subunits are identical in the region used for the antigen, it is expected that both isoforms will be detected by the antibody.
Human
Videos
Documents

For Research Use Only