missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540643
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GGGGGGGGGA GGGGGGMQPG SAAVTSGAYR GDPSSVKMVQ SDFMQGAMAA Sequence homology: Human: 100%; Mouse: 100%; Rat: 100%
POU3F3 Polyclonal antibody specifically detects POU3F3 in Human samples. It is validated for Western Blot.Specifications
POU3F3 | |
Polyclonal | |
Unconjugated | |
POU3F3 | |
8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5455 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P20264 | |
POU3F3 | |
Synthetic peptide directed towards the N-terminal of human POU3F3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |