missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519305
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with human based on sequence homology.
POU3F3 Polyclonal antibody specifically detects POU3F3 in Mouse samples. It is validated for Western Blot.Specifications
POU3F3 | |
Polyclonal | |
Unconjugated | |
POU3F3 | |
8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
18993 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P31361 | |
POU3F3 | |
Synthetic peptide sequence (HMLSHAHQWVTAL) corresponding to the internal amino acids of POU3F3 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |