missing translation for 'onlineSavingsMsg'
Learn More

POU3F2 Monoclonal Antibody (6H12), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA517153

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15338100

  • £394.00 / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MA5-17153 targets POU3F2 in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17153 immunogen is purified recombinant fragment of human POU3F2 (amino acids: 214-362) expressed in E. Coli. MA5-17153 detects POU3F2 which has a predicted molecular weight of approximately 46.9kDa.

POU3F2 Monoclonal antibody specifically detects POU3F2 in Human samples. It is validated for ELISA, Western Blot.
Specifications

Specifications

POU3F2
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P20265
POU3F2
Purified recombinant fragment of human POU3F2 (amino acids: 214-362) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Western Blot
6H12
Unconjugated
POU3F2
7-Oct; 9430075J19Rik; A230098E07Rik; Brain-2; brain-2 class III POU-domain protein; brain-specific homeobox/POU domain protein 2; BRN2; Brn-2; Nervous system-specific octamer-binding transcription factor N-Oct-3; N-Oct3; OCT7; Oct-7; Octamer-binding protein 7; octamer-binding transcription factor 7; Otf7; OTF-7; POU class 3 homeobox 2; POU domain, class 3, transcription factor 2; POU3F2; POUF3; protein Brn-2; Unknown (protein for MGC:137739)
Mouse
Protein G
RUO
5454
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers