missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519303
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology.
PLA1 Polyclonal antibody specifically detects PLA1 in Human samples. It is validated for Western Blot.Specifications
PLA1 | |
Polyclonal | |
Unconjugated | |
POU2F3 | |
11-Oct; Epoc-1; FLJ40063; LOC553427 protein; MGC126698; Oct11; oct-11; Oct-11a; octamer-binding protein 11; Octamer-binding transcription factor 11; OTF11; OTF-11; PLA1; PLA-1; POU class 2 homeobox 3; POU domain transcription factor OCT11a; POU domain, class 2, transcription factor 3; POU transcription factor; POU2F3; Rov 1 pou-homeodomain protein; Skin; Skin-1a; Skn-1a; Skn-li; Transcription factor PLA-1; transcription factor Skn-1; zgc:136377; zgc:158534 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
25833 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9UKI9 | |
POU2F3 | |
Synthetic peptide sequence (EPHLEASQHLPVPKH) corresponding to the internal amino acids of POU2F3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |