missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM123 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129811
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-29811 detects Porimin from bovine, mouse, human, rat samples.
TMEM123 Polyclonal antibody specifically detects TMEM123 in Human, Mouse samples. It is validated for Western Blot.Specifications
TMEM123 | |
Polyclonal | |
Unconjugated | |
TMEM123 | |
2310075C12Rik; KCT3; KCT-3; keratinocytes associated transmembrane protein 3; keratinocytes-associated transmembrane protein 3; Porimin; PORMIN; pro oncosis receptor inducing membrane injury; pro-oncosis receptor inducing membrane injury; PSEC0111; RGD1305625; serine/threonine-rich receptor; TMEM123; Transmembrane protein 123; UNQ641/PRO1271 | |
Rabbit | |
Protein G | |
RUO | |
114908, 71929 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q8N131, Q91Z22 | |
TMEM123 | |
synthetic peptide SRRGIRYRTIDEHDAI, corresponding to amino acids 173-188 of Human Porimin | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |