Learn More
Porimin Mouse anti-Human, Clone: 297617, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3010
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469835
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Porimin Monoclonal antibody MAB3010 Specifically detects Porimin in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
Porimin
297617
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TMEM123
Chinese hamster ovary cell line CHO-derived recombinant human Porimin His27-Ser149 Accession # AAG21694
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
114908
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
AAG21694
KCT3, KCT-3, KCT3keratinocytes associated transmembrane protein 3, Keratinocytes-associated transmembrane protein 3, PORIMINporimin, pro oncosis receptor inducing membrane injury, Pro-oncosis receptor inducing membrane injury, serine/threonine-rich receptor, TMEM123, transmembrane protein 123PORMIN
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Porimin in direct ELISAs and Western blots.