Learn More
Goat anti-Pig IgG Fc Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184628
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-84628 shows no cross-reactivity with other porcine immunoglobulin classes in immunoelectrophoresis. This antibody may react with IgG from other species. Goat anti Pig IgG (Fc) antibody recognizes the Fc region of the porcine IgG heavy chains and shows no cross - reactivity with other porcine immunoglobulin classes as evaluated by immunoelectrophoresis.
Pig IgG Fc Polyclonal antibody specifically detects Pig IgG Fc in Porcine samples. It is validated for ELISA.Specifications
Pig IgG Fc | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Porcine | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% Pro-Clean 400 | |
Purified porcine IgG. | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |