missing translation for 'onlineSavingsMsg'
Learn More

CD31/PECAM-1 Goat anti-Porcine, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3387

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428144

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD31/PECAM-1 Polyclonal antibody AF3387 Specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Western Blotting
Specifications

Specifications

CD31/PECAM-1
LYOPH
Western Blot 0.1 ug/mL
Q95242
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects porcine CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human CD31 and recombinant mouse CD31 is observed.
Porcine
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PECAM1
Mouse myeloma cell line NS0-derived recombinant porcine CD31/PECAM-1 Gln28-Lys602 Accession # Q95242
Immunogen affinity purified
RUO
Primary
5175
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only