Learn More
Poliovirus Type 1 Monoclonal Antibody (9B4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen HYB2951702
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
HYB 295-17-02 is detects Polio component type I. HYB 295-17-02 has been successfully used in ELISA procedures. HYB 295-17-02 reacts with Polio component type I. Reaction is seen in ELISA with Polio component type I directly coated onto the microtiter well. The HYB 295-17-02 immunogen is Polio component type I. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Poliovirus is a single-stranded RNA virus that belongs to the Picornavirus family, characterized by their small RNA genome (7.2bk to 8.5 kb). Poliovirus is a enterovirus which primarily infects the gastroentestinal assiciated lymphoid tissue (GALT) and can eventually spread to the central nervous system, causing paralysis. Mortality, though very rare, is usually due to respiratory failure by paralysis of the intercostal muscles and diaphragm. There are three separate types of the virus: Type 1, Type 2 and Type 3.Specifications
Poliovirus Type 1 | |
Monoclonal | |
1.02 mg/mL | |
PV-1 | |
Poliovirus type I. | |
200 μL | |
Primary | |
4° C, store in dark | |
Liquid |
ELISA | |
9B4 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2b κ |