missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PODXL Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518729
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
PODXL Polyclonal antibody specifically detects PODXL in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PODXL | |
Polyclonal | |
Unconjugated | |
PODXL | |
AW121214; GCTM-2 antigen; Gp200; Ly102; MGC138240; PC; PCLP; PCLP antibody; Pclp1; PCLP-1; PCLP1 antibody; PODLX2; podocalyxin; podocalyxin like; podocalyxin-like; podocalyxin-like protein 1; podolyxin; Podxl | |
Goat | |
Antigen affinity chromatography | |
RUO | |
192181, 27205, 5420 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O00592, Q9R0M4, Q9WTQ2 | |
PODXL | |
Synthetic peptide sequence (DNLTKDDLDEEEDTH) corresponding to the C-terminus amino acids of PODXL | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |