missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAAR5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521919
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, HeLa, HepG2, mouse muscle. Predicted reactivity: Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TAAR5 Polyclonal antibody specifically detects TAAR5 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
TAAR5 | |
Polyclonal | |
Unconjugated | |
Taar5 | |
Gm227; hTaar5; MGC138414; MGC138416; mTaar5; PNR; Putative neurotransmitter receptor; RP11-295F4.5; TAAR5; TaR-5; trace amine associated receptor 5; trace amine receptor 5; trace amine-associated receptor 5; Trimethylamine receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
215854, 9038 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O14804, Q5QD14 | |
Taar5 | |
Synthetic peptide corresponding to a region within amino acids 273 and 337 of TAAR5 (Uniprot ID#O14804) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |