Learn More
PNPase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522397
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (92%), Rat (93%), Pig (93%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PNPase Polyclonal antibody specifically detects PNPase in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
PNPase | |
Polyclonal | |
Unconjugated | |
PNPT1 | |
1200003F12Rik; 3'-5' RNA exonuclease OLD35; COXPD13; DFNB70; DKFZp762K1914; old 35; Old35; old-35; PNPase; PNPase 1; PNPase old-35; Pnpt1; Pnptl1; polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; polyribonucleotide nucleotidyltransferase 1; polyribonucleotide nucleotidyltransferase 1, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
87178 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TCS8 | |
PNPT1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 783 of PNPase (Uniprot ID#Q8TCS8) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |