Learn More
PNAS4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542033
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SCLPKEWLTP AALQSSVSQE LQDELEEAED AAASASVAST AAGSRPGRHT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
PNAS4 Polyclonal antibody specifically detects PNAS4 in Human samples. It is validated for Western Blot.Specifications
PNAS4 | |
Polyclonal | |
Unconjugated | |
DESI2 | |
5830417C01Rik; C1orf121; cb238; CGI-146; DESI; DESI1; desi2; DeSI-2; desumoylating isopeptidase 1; desumoylating isopeptidase 2; Deubiquitinase DESI2; Fam152a; family with sequence similarity 152, member A; fi12d12; hypothetical protein LOC445138; pnas4; PNAS-4; PPPDE peptidase domain containing 1; PPPDE peptidase domain-containing protein 1; Pppde1; Protein FAM152A; sb:cb238; si:ch211-132e22.3; wu:fi12d12; zgc:100860 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51029 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BSY9 | |
DESI2 | |
Synthetic peptide directed towards the C-terminal of human DESI2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |