Learn More
PMCA1 ATPase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1914
Code : LE
Additional Details : Weight : 1.24740kg
Description
PA1-914 detects calcium pump of the plasma membrane 1a (PMCA1a ATPase) and 1b from rat, human and porcine tissues. PA1-914 has been successfully used in Western blot, immunohistochemistry (paraffin and frozen), and ELISA procedures. By Western blot, this antibody detects ∽134 kDa and ∽130 kDa proteins representing PMCA1a and 1b ATPase from rat, human and porcine samples. The PA1-914 immunogen is a synthetic peptide corresponding to the residues A(5) N N S V A Y S G V K N S I K E A N(22) of rat PMCA1 ATPase.
PMCA1 ATPase Polyclonal antibody specifically detects PMCA1 ATPase in Human, Porcine, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
PMCA1 ATPase | |
Polyclonal | |
Unconjugated | |
ATP2B1 | |
2810442I22Rik; Atp2b1; ATPase plasma membrane Ca2+ transporting 1; ATPase, Ca++ transporting, plasma membrane 1; E130111D10Rik; plasma membrane Ca2+ pump (PMCA1b); plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium-transporting ATPase 1; PMCA1; Pmca1a; Pmca1b; Pmca1c; PMCA1kb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29598, 397636, 490 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P11505, P20020, P23220 | |
ATP2B1 | |
Synthetic peptide corresponding to the residues A(5) N N S V A Y S G V K N S I K E A N(22) of rat PMCA1 ATPase. | |
100 μg | |
Primary | |
Human, Porcine, Rat | |
Antibody | |
IgG |