missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGF Monoclonal Antibody (24G3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1188
Code : NEW
Additional Details : Weight : 0.00900kg
Description
By Western blot, MA1-188 detects both 13kDa monomer and 25kDa homodimer.
PGF Monoclonal antibody specifically detects PGF in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
PGF | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P49763 | |
PGF | |
Human recombinant Placental Growth Factor-1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
24G3 | |
Unconjugated | |
PGF | |
AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760 | |
Mouse | |
Protein A | |
RUO | |
5228 | |
-20°C | |
Liquid |