missing translation for 'onlineSavingsMsg'
Learn More

Plexin A1 Rat anti-Mouse, Clone: 408305, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4309

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460506

  • £264.01 / 100µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Plexin A1 Monoclonal antibody MAB4309 Specifically detects Plexin A1 in Mouse samples. It is validated for Immunohistochemistry
Specifications

Specifications

Plexin A1
408305
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PLXNA1
E. coli-derived recombinant mouse Plexin A1, Leu49-Gly165, Accession # P70206
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5361
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
P70206
NOV, NOVP, plexin A1, PLEXIN-A1, PLXN1, PLXNA1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Plexin A1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 10% cross-reactivity with recombinant mouse (rm) Plexin A3 is observed and no cross-reactivity with rmPlexin A2, recombinant human (rh) Plexin B1, rhPlexin C1, or rhPlexin D1 is observed.
Mouse
Videos

For Research Use Only