Learn More
Plasma Kallikrein/KLKB1 Rat anti-Mouse, Clone: 337801, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2498
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459485
  • £301.92 / 100µg
Estimated Shipment
10-08-2020
Description
Description
Plasma Kallikrein/KLKB1 Monoclonal antibody MAB2498 Specifically detects Plasma Kallikrein/KLKB1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Plasma Kallikrein/KLKB1
337801
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KLKB1
Mouse myeloma cell line NS0-derived recombinant mouse Kallikrein/KLKB1 Gly20-Ala638 Accession # NP_032481
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3818
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_032481
EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, KLKB1, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Plasma Kallikrein/KLKB1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Kallikrein/KLKB1, rhKallikrein 1, 3, 4, 5, 7, 8, 10, 11, 12, 13, 14, 15, recombinant mouse HGFA, rhFactor VI, rhFactor X, rhFactor XI, rhThrombin, or rhuPA is observed.