Learn More
PKC alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517551
Code : LE
Additional Details : Weight : 0.01000kg
Description
Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay. It is not recommended to aliquot this antibody. This antibody is not cross-reactive with endogenous levels of other PKC isoforms.
PKC alpha Polyclonal antibody specifically detects PKC alpha in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
PKC alpha | |
Polyclonal | |
Unconjugated | |
PRKCA | |
AAG6; aging-associated gene 6; AI875142; EC 2.7.11.13; FHC; FTH; FTHL6; im:7139045; kinase PKC-alpha; KPCA; MGC104426; PIG15; PKC III; Pkca; PKC-A; PKC-alpha; PKC-III; PKRCA; PLIF; PRKACA; Prkca; prkcaa; protein kinase C; protein kinase C alpha; protein kinase C alpha type; protein kinase C alpha type; protein kinase C, alpha, a; protein kinase C, alpha; protein kinase C, alpha, a; protein kinase c-alpha; si:dkey-8l19.1; Unknown (protein for IMAGE:7139045); unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18750, 24680, 5578 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
11 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P05696, P17252, P20444 | |
PRKCA | |
Synthetic peptide corresponding to human PKCa | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |