missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGW Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537196
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 57 kDa. Purity is >95% by SDS-PAGE.
PIGW Polyclonal antibody specifically detects PIGW in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PIGW | |
Polyclonal | |
Unconjugated | |
PIGW | |
2610044A17Rik; Gwt1; HPMRS5; phosphatidylinositol glycan anchor biosynthesis class W; phosphatidylinositol glycan anchor biosynthesis, class W; phosphatidylinositol glycan, class W; phosphatidylinositol-glycan biosynthesis class W protein; Pigw; PIG-W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284098, 378774, 70325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7TSN4, Q7Z7B1, Q8C398 | |
PIGW | |
A synthetic peptide corresponding to residues in human PIGW | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |