missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535340
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PIGR Polyclonal antibody specifically detects PIGR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
PIGR | |
Polyclonal | |
Unconjugated | |
Pigr | |
hepatocellular carcinoma associated protein TB6; hepatocellular carcinoma-associated protein TB6; phosphatidylinositol glycan, class R; Pigr; Poly Ig receptor; poly-Ig receptor; polymeric immunoglobulin receptor; Secretory component; tb6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5284 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P01833 | |
Pigr | |
KLH conjugated synthetic peptide between 646-672 amino acids from the C-terminal region of human PIGR | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |