missing translation for 'onlineSavingsMsg'
Learn More

PIG3, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA112547

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11566022

  • £274.00 / 100µg


Estimated Shipment
24-05-2018
Add to basket

Specifications

Specifications

PIG3
Polyclonal
PBS with 0.08% sodium azide
Q53FA7
p53 Inducible Gene 3
Rabbit
IgG
100μg
Antibody
Polyclonal
Human
Western Blot
Unconjugated
TP53I3
Purified
TP53I3
Full-length recombinant PIG-3 protein.
Ammonium sulfate precipitation
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
9540
Videos