missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphotyrosine Mouse, HRP, Clone: PY20, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610011
Additional Details : Weight : 0.01000kg
Description
Phosphorylation of specific tyrosine residues is the result of of activation or stimulation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be autophosphorylated kinases or certain cellular protein substrates that are regulated in oncogenesis or cell growth. Antibodies to phosphotyrosine provide one of the best tools for the detection and characterization of phosphotyrosine proteins.Specifications
Phosphotyrosine | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA and glycerol. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Western Blot | |
PY20 | |
HRP | |
Mouse | |
50 μg | |
Cell Biology | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
IgG2b |
For Research Use Only.