missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-THRB (Ser142) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539818
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Thyroid hormone receptors (TRs) are ligand dependent members of the steroid/retinoic acid superfamily of transcription factors. The two genes encoding TRs identified to date, TR alpha and TR beta, have been mapped to human chromosomes 17 and 3, respectively. TRs bind to thyroid hormone response elements (TREs) with half site binding motifs in the orientation of palindromes, direct repeats, or inverted palindromes. The affinities of binding are both variable and influenced differentially by 3,5,3′-triiodo-L-thyronine (T-3).Specifications
Phospho-THRB (Ser142) | |
Polyclonal | |
Unconjugated | |
THRB | |
TR-B1C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2, nuclear receptor subfamily 1 group A member 2, oncogene ERBA2, thyroid hormone nuclear receptor beta variant 1, thyroid hormone receptor beta, thyroid hormone receptor, beta erythroblastic leukemia viral v-erb-a oncogene homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21834, 24831, 7068 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P10828, P37242, P18113 | |
THRB | |
A synthetic phosphopeptide derived from human TR-beta 1 around the phosphorylation site of Ser142 (H-P-SP-Y-S) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |