missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT6 (Tyr641) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517902
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the corresponding phospho-tyrosine residues of other Stat proteins.
Phospho-STAT6 (Tyr641) Polyclonal antibody specifically detects Phospho-STAT6 (Tyr641) in Bovine, Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Phospho-STAT6 (Tyr641) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P42226 | |
STAT6 | |
Synthetic phosphopeptide corresponding to residues surrounding pTyr641 of human Stat6 | |
100 μL | |
Primary | |
Bovine, Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
STAT6 | |
12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
353105, 6778 | |
-20°C | |
Liquid |