missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT3 (Tyr705) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44380G
Code : 50
Additional Details : Weight : 0.05000kg
Description
Phospho-STAT3 (Tyr705) Polyclonal antibody specifically detects Phospho-STAT3 (Tyr705) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-STAT3 (Tyr705) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3 | |
P40763, P42227, P52631 | |
STAT3 | |
The antiserum was produced against a chemically synthesized phosphorylated peptide derived from the region of human STAT3 that contains tyrosine 705. The sequence is conserved in mouse and rat. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
STAT3 | |
1110034C02Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; APRF; AW109958; DNA-binding protein APRF; FLJ20882; HIES; MGC16063; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); signal transducer; activator of transcription; acute-phase response factor; signal transduction and activation of transcription 3; STAT; STAT3; STAT-3; stat3 protein; STAT3b1; STAT3b2; transcription factor; Unknown (protein for MGC:128731); wu:fc15d02; wu:fl59g06; z-Stat3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20848, 25125, 6774 | |
-20°C | |
Liquid |