missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-POLR2A (Ser1619) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538091
Code : NEW
Additional Details : Weight : 0.00900kg
Description
RPB1 is a subunit of RNA polymerase II which catalyses transcription of RNA from DNA. RPB1 forms the polymerase active center together with the second largest subunit. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB1 is part of the core element with the central large cleft, the clamp element that moves to open and close the cleft and the jaws that are thought to grab the incoming DNA template.Specifications
Phospho-POLR2A (Ser1619) | |
Polyclonal | |
Unconjugated | |
POLR2A | |
RPO2; RPOL2,POLR2 ALPHA-AMANITIN RESISTANCE,POLYMERASE II, RNA, SUBUNIT A,RNA POLYMERASE II, 220-KD SUBUNIT,RNA POLYMERASE II, LARGE SUBUNIT,RPB1, S. CEREVISIAE, HOMOLOG OF polymerase (RNA) II (DNA directed) polypeptide A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20020, 363633, 5430 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P24928, P08775 | |
POLR2A | |
A synthetic phosphopeptide derived from human POLR2A around the phosphorylation site of Ser1619 (P-T-SP-P-S) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |