Learn More
Phospho-c-Raf (Ser301) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14639
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, chicken, mouse, non-human primate and Xenopus based on 100% sequence homology. This antibody is specific for the ∽74 kDa Raf-1 protein phosphorylated at Ser301. Immunolabeling can be blocked by lambda-phosphatase treatment.
Phospho-c-Raf (Ser301) Polyclonal antibody specifically detects Phospho-c-Raf (Ser301) in Human, Rat samples. It is validated for Western Blot.Specifications
Phospho-c-Raf (Ser301) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P04049, P11345 | |
Raf1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser301 of rat RAF1 conjugated to KLH | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Raf1 | |
6430402F14Rik; AA990557; BB129353; CMD1NN; CRAF; C-RAF; C-Raf proto-oncogene, serine/threonine kinase; Craf1; Craf1 transforming; D830050J10Rik; EC 2.7.11.1; kinase Raf1; murine leukemia viral (v-raf-1) oncogene homolog 1 (3611-MSV); murine sarcoma 3611 oncogene 1; NS5; Oncogene RAF1; OTTHUMP00000207813; OTTHUMP00000209389; proto-oncogene c-RAF; RAF; raf proto-oncogene serine/threonine protein kinase; RAF proto-oncogene serine/threonine-protein kinase; RAF1; Raf-1; Raf-1 proto-oncogene, serine/threonine kinase; sarcoma 3611 oncogene; v-Raf; v-raf-1 leukemia viral oncogene 1; vraf1 murine le; v-raf-1 murine leukemia viral oncogene homolog 1; v-raf-1 murine leukemia viral oncogene-like protein 1; v-raf-leukemia viral oncogene 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
24703, 5894 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |