missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RSK1 (Thr359, Ser363) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538309
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The p90 ribosomal S6 kinases (RSKs) comprise a family of serine/threonine kinases that lie at the terminus of the ERK pathway. In humans, the RSK family consists of four isoforms (RSK1 to-4). RSK family members are unusual among serine/threonine kinases in that they contain two distinct kinase domains, both of which are catalytically functional. Theses kinase dimains are activated in a sequential manner by a series of phosphorylations. RSK regulates gene expression via association and phosphorylation of transcriptional regulators including c-Fos, estrogen receptor, NFkappaB/IkappaB alpha, cAMP-response element-binding protein (CREB). ERK activates the C-terminal kinase of RSK, leading to activation of the N-terminal kinase. Members of the RSK family are present in the cytoplasm as well as the nucleus. Addition of growth factor to the cells results in the activation of both cytosolic and nuclear RSK and the translocation of the cytosolic RSK into the nucleus upon activation. The activation and nuclear translocation of RSK result in phosphorylation and activation of transcription factors.Specifications
Phospho-RSK1 (Thr359, Ser363) | |
Polyclonal | |
Unconjugated | |
RPS6KA1 | |
HU-1, MAPKAPK1A, RSK, RSK1, 90 kDa ribosomal protein S6 kinase 1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, RSK-1, S6K-alpha 1, S6K-alpha-1, dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1), p90-RSK 1, p90RSK1, p90S6K, r, HU-2, MAPKAPK1C, RSK, RSK3, S6K-alpha, S6K-alpha2, p90-RSK3, pp90RSK3, 90 kDa ribosomal protein S6 kinase 2, MAP kinase-activated protein kinase 1c, MAPK-activated protein kinase 1c, MAPKAP kinase 1c, MAPKAPK-1c, RSK-3, S6K-alpha-2, p90-RSK 2, p90RSK2, ribosomal S6 kinase 3, ribosomal protein S6 kinase alpha 2, ribosomal protein S6 kin, RP11-393H10.3, CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2, 90 kDa ribosomal protein S6 kinase 3, MAP kinase-activated protein kinase 1b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20111, 6195, 81771 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15418, P18653, Q63531 | |
RPS6KA1 | |
A synthetic phosphopeptide derived from human P90RSK around the phosphorylation site of Thr359/Ser363 (TP-P-K-D-SP) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |