missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p70 S6 Kinase (Thr229) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538306
Code : NEW
Weitere Details : Weight : 0.00900kg
Beschreibung
The p70 S6 kinase (p70 Ribosomal Protein S6 Kinase, p70S6K) is a 70 kDa member of the ribosomal S6 kinase (RSK) family of serine/threonine kinases. p70 S6 kinase is predomitly localized in the cytoplasm, and is essential in growth factors regulated cell proliferation, pathways involving cell motility, such as metastases, the immune response, and tissue repair. p70 S6 kinase acts downstream of phosphoinositide (PI) 3-kinase, and its main physiological target is the S6 ribosomal protein, which is involved in upregulation of protein synthesis. The kinase activity of p70 S6 kinase leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding p70 S6 kinase and overexpression are seen in some breast cancer cell lines.Spezifikation
Phospho-p70 S6 Kinase (Thr229) | |
Polyclonal | |
Unconjugated | |
RPS6KB1 | |
Ribosomal protein S6 kinase I, S6K, S6K1, 70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase alpha, p70 S6K, p70-S6K, P70S6K, p70 S6 kinase alpha, Name: RPS6KB1, STK14A, p80 S6K1, p85 S6 kinase, p85 S6K, p85S6K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6198, 72508, 83840 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P23443, Q8BSK8, P67999 | |
RPS6KB1 | |
A synthetic phosphopeptide derived from human P70S6K around the phosphorylation site of Thr229 (T-H-TP-F-C) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |