missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p53 (Ser46) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517817
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with p53 phosphorylated at other sites.
Phospho-p53 (Ser46) Polyclonal antibody specifically detects Phospho-p53 (Ser46) in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Phospho-p53 (Ser46) | |
Polyclonal | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7157 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
20 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P04637 | |
Tp53 | |
Synthetic phosphopeptide corresponding to residues surrounding pSer46 of human p53 | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |