missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-p27 Kip1 (Thr157) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA512650
Code : LE
Additional Details : Weight : 0.01000kg
Description
Phospho-p27 Kip1 (Thr157) Polyclonal antibody specifically detects Phospho-p27 Kip1 (Thr157) in Human samples. It is validated for Dot Blot, Immunohistochemistry.Specifications
Phospho-p27 Kip1 (Thr157) | |
Polyclonal | |
Unconjugated | |
Cdkn1b | |
AA408329; AI843786; Cdki1b; CDKN1B; CDKN4; CDN1B; cyclin dependent kinase inhibitor 1B; cyclin-dependent kinase inhibitor 1B; Cyclin-dependent kinase inhibitor 1B (p27 Kip1); Cyclin-dependent kinase inhibitor 1B (p27, Kip1); cyclin-dependent kinase inhibitor p27; Cyclin-dependent kinase inhibitor p27 Kip1; cyclin-dependent kinase inhibitor p27/Kip1; KIP1; MEN1B; MEN4; p27; P27 CDKN1B; p27(KIP1); p27Kip1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P46527 | |
Cdkn1b | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T157 of human p27Kip1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |