missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MEK1 (Thr292) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44458G
Code : 50
Additional Details : Weight : 0.50000kg
Description
Phospho-MEK1 (Thr292) Polyclonal antibody specifically detects Phospho-MEK1 (Thr292) in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-MEK1 (Thr292) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P31938, Q01986, Q02750 | |
Map2k1 | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human MEK1 that contains threonine 292. The sequence is conserved in many species including mouse, rat, chimp, hamster and rabbit. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Map2k1 | |
CFC3; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase or Erk Kinase, Dual specificity mitogen-activated protein kinase kinase, involved in ras mediated vulval induction, LEThal LET-537 (42.8 kD) (mek-2); MAP kinase/Erk kinase 1; Map2k1; map2k1.L; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK 1; Mek1; MEK2; mek-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1 L homeolog; MKK (Thr292); MKK (Thr386); MKK1; MKK2; MP2K1; Prkmk1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); XELAEV_18018447mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170851, 26395, 5604 | |
-20°C | |
Liquid |