missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MCAK (Ser95) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538864
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Phospho-MCAK (Ser95) Polyclonal antibody specifically detects Phospho-MCAK (Ser95) in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Phospho-MCAK (Ser95) | |
Polyclonal | |
Unconjugated | |
KIF2C | |
4930402F02Rik; CT139; ESTM5; hypothetical protein; KIF2C; kinesin family member 2C; kinesin-like 6; kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; KNSL6; KRP2; MCAK; Mitotic centromere-associated kinesin; QtsA-16015; RP11-269F19.1; testis tissue sperm-binding protein Li 68n; X83316 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11004, 73804 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q922S8, Q99661 | |
KIF2C | |
A synthetic phosphopeptide derived from human KIF2C around the phosphorylation site of Ser95 (R-R-SP-V-N) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |