Learn More
Phospho-IGF1R beta (Tyr1135, Tyr1136) Recombinant Rabbit Monoclonal Antibody (15H10L8), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen 701067
Code : NEW
Additional Details : Weight : 0.00450kg
Description
Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Phospho-IGF1R beta (Tyr1135, Tyr1136) Recombinant Monoclonal antibody specifically detects Phospho-IGF1R beta (Tyr1135, Tyr1136) in Human samples. It is validated for Western Blot.Specifications
Phospho-IGF1R beta (Tyr1135, Tyr1136) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Phosphopeptide corresponding to amino acids 1131–1140 of human IGF1R | |
100 μg | |
Primary | |
Human | |
Liquid |
Western Blot | |
15H10L8 | |
Unconjugated | |
P08069 | |
Rabbit | |
Protein A | |
RUO | |
3480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |