missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-hnRNP C (Ser260) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538589
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Binds pre-mRNA and nucleates the assembly of 40S hnRNP particles. Single HNRNPC tetramers bind 230-240 nucleotides. Trimers of HNRNPC tetramers bind 700 nucleotides. May play a role in the early steps of spliceosome assembly and pre-mRNA splicing. Interacts with poly-U tracts in the 3′-UTR or 5′-UTR of mRNA and modulates the stability and the level of translation of bound mRNA molecules.Specifications
Phospho-hnRNP C (Ser260) | |
Polyclonal | |
Unconjugated | |
HNRNPC | |
Heterogeneous nuclear ribonucleoproteins C1/C2, hnRNP C1/C2, HNRNPC, HNRPC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15381, 3183 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P07910, Q9Z204 | |
HNRNPC | |
A synthetic phosphopeptide derived from human hnRNP C1/2 around the phosphorylation site of Ser260 (D-D-SP-A-E) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |