missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537817
Code : NEW
Additional Details : Weight : 0.00910kg
Description
A suggested positive control for Western blot is 293 cells.
Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) Polyclonal antibody specifically detects Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) in Human, Mouse samples. It is validated for Western Blot.Specifications
Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) | |
Polyclonal | |
Unconjugated | |
FOXO4 | |
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; AFX; AFX1; Afxh; AI876417; C76856; FKH1; FKHR; Fkhr1; Fkhr2; Fkhr3; FKHRL1; FKHRL1P2; fork head domain transcription factor AFX1; forkhead box O1; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box O3; forkhead box O3A; forkhead box O4; forkhead box protein O1; Forkhead box protein O1A; forkhead box protein O3; Forkhead box protein O4; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma; Forkhead in rhabdomyosarcoma-like 1; forkhead protein; forkhead protein 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; Foxo1; FOXO1A; FOXO2; FOXO3; Foxo3a; FOXO4; MGC120490; Mllt7; myeloid/lymphoid or mixed lineage-leukemia translocation to 7 homolog; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 7; RGD1561201 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2308, 2309, 4303, 54601, 56458, 56484 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43524, P98177, Q12778, Q9R1E0, Q9WVH3, Q9WVH4 | |
FOXO1, FOXO3, FOXO4 | |
Peptide sequence around phosphorylation site of threonine 24/32(S-C-T(p)-W-P) derived from Human FOXO1/3/4-pan. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |