missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-DAPP1 (Tyr139) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538468
Code : NEW
Additional Details : Weight : 0.00900kg
Description
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.Specifications
Phospho-DAPP1 (Tyr139) | |
Polyclonal | |
Unconjugated | |
DAPP1 | |
BAM32, B-cell adapter molecule of 32 kDa, b lymphocyte adapter protein Bam32, dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide, hDAPP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26377, 27071 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UN19, Q9QXT1 | |
DAPP1 | |
A synthetic phosphopeptide derived from human DAPP1 around the phosphorylation site of Tyr139 (S-I-YP-E-S) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |