missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-c-Jun (Ser73) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44292G
Code : 50
Additional Details : Weight : 0.01000kg
Description
Phospho-c-Jun (Ser73) Polyclonal antibody specifically detects Phospho-c-Jun (Ser73) in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Phospho-c-Jun (Ser73) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P05412, P05627 | |
JUN | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human c-Jun that contains serine 73. The sequence is conserved among many species including mouse, rat and chicken. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
JUN | |
Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
16476, 3725 | |
-20°C | |
Liquid |