missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Aurora B (Thr232) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538557
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Aurora kinases belong to a highly conserved family of mitotic serine/threonine kinases with three members identified among mammals: Aurora A, B, and C. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.Specifications
Phospho-Aurora B (Thr232) | |
Polyclonal | |
Unconjugated | |
AURKB | |
Aurora kinase B, Aurora 1, Aurora- and IPL1-like midbody-associated protein 1, AIM-1, Aurora/IPL1-related kinase 2, ARK-2, Aurora-related kinase 2, STK-1, Serine/threonine-protein kinase 12, Serine/threonine-protein kinase 5,Serine/threonine-protein kinase aurora-B, AURKB, AIK2, AIM1, AIRK2, ARK2, STK1, STK12, STK5, Aurora kinase C, Aurora 3, Aurora/IPL1-related kinase 3, ARK-3, Aurora-related kinase 3, Aurora/IPL1/Eg2 protein 2, Serine/threonine-protein kinase 13, Serine/threonine-protein kinase aurora-C, AURKC, AIE2, AIK3, AIRK3, ARK3, STK13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114592, 20877, 9212 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96GD4, O70126, O55099 | |
AURKB | |
A synthetic phosphopeptide derived from human AurB around the phosphorylation site of Thr232 (R-K-TP-M-C) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |