missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGBD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541301
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AESDSDDPSY APKDDSPDEV PSTFTVQQPP PSRRRKMTKI LCKWKKADLT Sequence homology: Human: 100%
PGBD3 Polyclonal antibody specifically detects PGBD3 in Human samples. It is validated for Western Blot.Specifications
PGBD3 | |
Polyclonal | |
Unconjugated | |
PGBD3 | |
4732403I04; ATP-dependent helicase ERCC6; C130058G22Rik; Cockayne syndrome protein CSB; CS group B correcting; Csb; DNA excision repair protein ERCC-6; Ercc6; excision repair cross-complementation group 6; excision repair cross-complementing rodent repair deficiency, complementation group 6; FLJ90201; PGBD3; piggyBac transposable element derived 3; piggyBac transposable element-derived protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
267004 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N328 | |
PGBD3 | |
Synthetic peptide directed towards the N-terminal of human PGBD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |