Learn More
PGAM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521752
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T. Predicted reactivity: Xenopus laevis (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PGAM1 Polyclonal antibody specifically detects PGAM1 in Human samples. It is validated for Western Blot.Specifications
PGAM1 | |
Polyclonal | |
Unconjugated | |
PGAM1 | |
2310050F24Rik; BPG-dependent PGAM 1; CDABP0006; epididymis secretory protein Li 35; HEL-S-35; PGAM1; Pgam-1; PGAMA; PGAM-B; Pgmut; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase isozyme B; RP11-452K12.8; similar to phosphoglycerate mutase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5223 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.97 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P18669 | |
PGAM1 | |
Synthetic peptide corresponding to a region within amino acids 191 and 254 of PGAM1 (Uniprot ID#P18669) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |