Learn More
Periostin/OSF-2 Rat anti-Mouse, Clone: 345616, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2955
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469805
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Periostin/OSF-2 Monoclonal antibody MAB2955 Specifically detects Periostin/OSF-2 in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
Periostin/OSF-2
345616
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
NP_056599
Fasciclin I-like, MGC119510, MGC119511, OSF2, OSF-2, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1, POSTN, TRIF52
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Periostin/OSF-2 in ELISAs.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
POSTN
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Periostin/OSF-2 isoform 2 Asn24-Gln811 Accession # NP_056599
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10631
Reconstitute at 0.5 mg/mL in sterile PBS.