Learn More
PER1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1524
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-524 detects PER1 from human and mouse tissues as well as recombinant mouse PER1. PA1-524 has been sucessfully used in Western blot, immunofluoresence and immunohistochemistry procedures. By Western blot, this antibody detects an ∽136 kDa protein representing overexpressed recombinant mouse PER1 protein. Immunofluorescence staining of PER1 in mouse testes with PA1-524 results in cytoplasmic staining. PA1-524 immunizing peptide corresponds to amino acid residues 39-51 from mouse PER1. PA1-524 immunizing peptide (Cat. # PEP-076) is available for use in neutralization and control experiments.
PER1 Polyclonal antibody specifically detects PER1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
PER1 | |
Polyclonal | |
Unconjugated | |
PER1 | |
circadian clock protein PERIOD 1; Circadian pacemaker protein Rigui; hPER; hPER1; KIAA0482; mPer1; m-rigui; PER; PER1; per1 {ECO:0000312; period 1; period circadian clock 1; period circadian protein homolog 1; period homolog 1; Period, drosophila, homolog of; RGD:727863}; RIGUI; rPER1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18626, 5187 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O15534, O35973 | |
PER1 | |
Synthetic Peptide: P(39) S L A D D T D A N S N G(51) C | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |