missing translation for 'onlineSavingsMsg'
Learn More

Pentraxin 2/SAP Rat anti-Mouse, Clone: 300103, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2558

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459535

  • £264.01 / 100µg


Estimated Shipment
30-10-2018
Add to basket

Description

Description

Pentraxin 2/SAP Monoclonal antibody MAB2558 Specifically detects Pentraxin 2/SAP in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

Pentraxin 2/SAP
300103
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
APCS
Mouse myeloma cell line NS0-derived recombinant mouse Pentraxin 2/SAP, Gln21-Asp224, Accession # P12246
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
325
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P12246
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Pentraxin 2/SAP in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant rat Pentraxin 2/SAP is observed. In Western blots, 100% cross-reactivity with recombinant human (rh) Pentraxin 2/SAP is observed, but no cross-reactivity with rhPentraxin 2/SAP in direct ELISAs is observed.
Mouse
Videos

For Research Use Only