Learn More
Pentraxin 2/SAP Mouse anti-Rat, Clone: 273902, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB18951
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469005
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Pentraxin 2/SAP Monoclonal antibody MAB18951 Specifically detects Pentraxin 2/SAP in Rat samples. It is validated for Blocking (Receptor-ligand)
Specifications
Specifications
Pentraxin 2/SAP
273902
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
APCS
Mouse myeloma cell line NS0-derived recombinant rat Pentraxin 2/SAP Gln21-Ser228 (predicted) Accession # P23680
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
325
Reconstitute at 0.5 mg/mL in sterile PBS.
Blockade of Receptor-ligand Interaction
LYOPH
Blockade of Receptor-ligand Interaction
P23680
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat Pentraxin 2/SAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Pentraxin 2 or recombinant mouse Pentraxin 2 is observed.
Rat