missing translation for 'onlineSavingsMsg'
Learn More

PDGF R beta Rat anti-Mouse, Clone: 182106, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1042

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488275

  • £353.77 / 500µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

PDGF R beta Monoclonal antibody MAB1042 Specifically detects PDGF R beta in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

PDGF R beta
182106
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PDGFRB
Mouse myeloma cell line NS0-derived recombinant mouse PDGF R beta , Leu32-Lys530, Accession # P05622
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5159
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P05622
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGFRB, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Rat
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse PDGF R beta in direct ELISAs and Western blots. In direct ELISAs, approximately 10-20% cross-reactivity with recombinant human PDGF R beta is observed, but no cross-reactivity with recombinant mouse (rm) PDGF R alpha , rmFlt-3, rmSCF R, or rmLymphotoxin beta R is observed.
Mouse
Videos

For Research Use Only