missing translation for 'onlineSavingsMsg'
Learn More

PDGF R alpha Rat anti-Mouse, Clone: 189208, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1062

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458295

  • £353.77 / 500µg


Estimated Shipment
27-11-2018
Add to basket

Description

Description

PDGF R alpha Monoclonal antibody MAB1062 Specifically detects PDGF R alpha in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

PDGF R alpha
189208
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
PDGFRA
Mouse myeloma cell line NS0-derived recombinant mouse PDGF R alpha , Leu25-Glu524 (Asp65Glu, Gly439Ala, Thr440Ala), predicted, Accession # P26618
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5156
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P26618
alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse PDGF R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, 5-20% cross-reactivity with recombinant human PDGF R alpha is observed and no cross-reactivity with recombinant mouse PDGF R beta is observed.
Mouse
Videos

For Research Use Only